accessibility menu, dialog, popup

UserName

CAS RN 583-02-8

IUPAC Name: ethyl 4-(trifluoromethyl)benzoate
Synonyms: ethyl 4-(trifluoromethyl)benzoate
Molecular Weight (g/mol): 218.18
Molecular Formula: C10H9F3O2
InChi Key: ZQDADDSPMCHZPX-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(C=C1)C(F)(F)F
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)