missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-pyridineacetate, 98%

CAS: 54401-85-3 | C9H11NO2 | 165.192 g/mol

Supplier:  Thermo Scientific Chemicals A1455006

 View more versions of this product

Catalog No. AAA1455006


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl 4-pyridineacetate
18°C to 20°C
86°C to 88°C (1 mmHg)
C9H11NO2
MFCD00051830
123786
QVLJLWHOILVHJJ-UHFFFAOYSA-N
ethyl 2-pyridin-4-ylacetate
736321
98%
54401-85-3
1.08
>110°C (230°F)
1.5
5 g
ethyl 4-pyridylacetate, ethyl 2-pyridin-4-yl acetate, ethyl pyridine-4-acetate, ethyl pyridin-4-ylacetate, 4-pyridineacetic acid ethyl ester, 4-pyridineacetic acid, ethyl ester, ethyl 2-4-pyridyl acetate, 4-pyridylacetic acid ethyl ester, ethyl 4-pyridineacetate, ethyl-4-pyridylacetate
CCOC(=O)CC1=CC=NC=C1
165.192
165.19

Chemical Identifiers

54401-85-3
165.192
QVLJLWHOILVHJJ-UHFFFAOYSA-N
736321
CCOC(=O)CC1=CC=NC=C1
C9H11NO2
MFCD00051830
ethyl 4-pyridylacetate, ethyl 2-pyridin-4-yl acetate, ethyl pyridine-4-acetate, ethyl pyridin-4-ylacetate, 4-pyridineacetic acid ethyl ester, 4-pyridineacetic acid, ethyl ester, ethyl 2-4-pyridyl acetate, 4-pyridylacetic acid ethyl ester, ethyl 4-pyridineacetate, ethyl-4-pyridylacetate
ethyl 2-pyridin-4-ylacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 259-150-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only