accessibility menu, dialog, popup

UserName

H3COON

CAS RN 54401-85-3

IUPAC Name: ethyl 2-(pyridin-4-yl)acetate
Synonyms: ethyl 2-(pyridin-4-yl)acetate
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: QVLJLWHOILVHJJ-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=NC=C1
Boiling Point 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)