missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-pyridineacetate, 98%

Catalog No. p-7021987
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1455003 1 g
AAA1455006 5 g
AAA1455014 25 g
AAA1455022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1455003 Supplier Thermo Scientific Chemicals Supplier No. A1455003
Only null left

CAS: 54401-85-3 | C9H11NO2 | 165.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54401-85-3
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00051830
InChI Key QVLJLWHOILVHJJ-UHFFFAOYSA-N
Synonym ethyl 4-pyridylacetate, ethyl 2-pyridin-4-yl acetate, ethyl pyridine-4-acetate, ethyl pyridin-4-ylacetate, 4-pyridineacetic acid ethyl ester, 4-pyridineacetic acid, ethyl ester, ethyl 2-4-pyridyl acetate, 4-pyridylacetic acid ethyl ester, ethyl 4-pyridineacetate, ethyl-4-pyridylacetate
PubChem CID 736321
IUPAC Name ethyl 2-pyridin-4-ylacetate
SMILES CCOC(=O)CC1=CC=NC=C1

Specifications

Melting Point 18°C to 20°C
Density 1.08
Boiling Point 86°C to 88°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5
Quantity 1 g
Beilstein 123786
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material Ethyl 4-pyridineacetate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 259-150-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only