missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Nitropyrrole-2-carboxylate 98.0+%, TCI America™

Catalog No. N05101G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05101G 1 g
N05105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05101G Supplier TCI America Supplier No. N05101G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5930-92-7
Molecular Formula C7H8N2O4
Molecular Weight (g/mol) 184.151
MDL Number MFCD00059927
InChI Key PEORWHVRWXGKMS-UHFFFAOYSA-N
Synonym 4-Nitropyrrole-2-carboxylic Acid Ethyl Ester
PubChem CID 242169
IUPAC Name ethyl 4-nitro-1H-pyrrole-2-carboxylate
SMILES CCOC(=O)C1=CC(=CN1)[N+](=O)[O-]

Specifications

Melting Point 174°C
Color White-Yellow
Quantity 1 g
UN Number 1325
Formula Weight 184.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Nitropyrrole-2-carboxylate