accessibility menu, dialog, popup

UserName

CH3OONHNOO

CAS RN 5930-92-7

IUPAC Name: ethyl 4-nitro-1H-pyrrole-2-carboxylate
Synonyms: ethyl 4-nitro-1H-pyrrole-2-carboxylate
Molecular Weight (g/mol): 184.15
Molecular Formula: C7H8N2O4
InChi Key: PEORWHVRWXGKMS-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(=CN1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results