missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-Methoxyphenylacetate 98.0+%, TCI America™
$162.17 - $1606.81
Chemical Identifiers
| CAS | 14062-18-1 |
|---|---|
| Molecular Formula | C11H14O3 |
| Molecular Weight (g/mol) | 194.23 |
| MDL Number | MFCD00040760 |
| InChI Key | DOCCDOCIYYDLGJ-UHFFFAOYSA-N |
| Synonym | ethyl 2-4-methoxyphenyl acetate, ethyl 4-methoxyphenylacetate, 4-methoxyphenylacetic acid ethyl ester, benzeneacetic acid, 4-methoxy-, ethyl ester, ethyl p-methoxyphenylacetate, ethyl 4-methoxyphenyl acetate, 4-methoxy-phenyl-acetic acid ethyl ester, acmc-1bnx2, homoanisic acid ethyl ester |
| PubChem CID | 84174 |
| IUPAC Name | ethyl 2-(4-methoxyphenyl)acetate |
| SMILES | CCOC(=O)CC1=CC=C(C=C1)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M079325ML
|
TCI America
M079325ML |
25 mL |
Each for $162.17
|
|
|||||
|
M0793500ML
|
TCI America
M0793500ML |
500 mL |
Each for $1,606.81
|
|
|||||
Chemical Identifiers
| 14062-18-1 | |
| 194.23 | |
| DOCCDOCIYYDLGJ-UHFFFAOYSA-N | |
| 84174 | |
| CCOC(=O)CC1=CC=C(C=C1)OC |
| C11H14O3 | |
| MFCD00040760 | |
| ethyl 2-4-methoxyphenyl acetate, ethyl 4-methoxyphenylacetate, 4-methoxyphenylacetic acid ethyl ester, benzeneacetic acid, 4-methoxy-, ethyl ester, ethyl p-methoxyphenylacetate, ethyl 4-methoxyphenyl acetate, 4-methoxy-phenyl-acetic acid ethyl ester, acmc-1bnx2, homoanisic acid ethyl ester | |
| ethyl 2-(4-methoxyphenyl)acetate |
Specifications
| 14062-18-1 | |
| 158°C | |
| MFCD00040760 | |
| 1993 | |
| DOCCDOCIYYDLGJ-UHFFFAOYSA-N | |
| ethyl 2-(4-methoxyphenyl)acetate | |
| 84174 | |
| ≥98.0% (GC) | |
| Ethyl 4-Methoxyphenylacetate |
| Yellow | |
| C11H14O3 | |
| 25 mL | |
| ethyl 2-4-methoxyphenyl acetate, ethyl 4-methoxyphenylacetate, 4-methoxyphenylacetic acid ethyl ester, benzeneacetic acid, 4-methoxy-, ethyl ester, ethyl p-methoxyphenylacetate, ethyl 4-methoxyphenyl acetate, 4-methoxy-phenyl-acetic acid ethyl ester, acmc-1bnx2, homoanisic acid ethyl ester | |
| CCOC(=O)CC1=CC=C(C=C1)OC | |
| 194.23 | |
| 194.23 | |
| Liquid |
Safety and Handling
TSCA : No