accessibility menu, dialog, popup

UserName

CAS RN 14062-18-1

H3COOOCH3

CAS RN 14062-18-1

IUPAC Name: ethyl 2-(4-methoxyphenyl)acetate
Synonyms: ethyl 2-(4-methoxyphenyl)acetate
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: DOCCDOCIYYDLGJ-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=C(OC)C=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)