missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-Methoxyphenylacetate 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 158°C |
| Quantity | 500 mL |
| UN Number | 1993 |
| Formula Weight | 194.23 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Ethyl 4-Methoxyphenylacetate |
Safety and Handling
| TSCA | No |