missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-iodobenzoate, 98%

CAS: 51934-41-9 | C9H9IO2 | 276.07 g/mol

Supplier:  Thermo Scientific Chemicals A1457018

 View more versions of this product

Catalog No. AAA1457018


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl 4-iodobenzoate
1.66
>110°C (230°F)
1.588
50 g
Light sensitive
YCBJOQUNPLTBGG-UHFFFAOYSA-N
ethyl 4-iodobenzoate
142891
98%
51934-41-9
283°C
C9H9IO2
MFCD00017344
2208568
p-iodobenzoic acid ethyl ester, 4-iodobenzoic acid ethyl ester, ethyl p-iodobenzoate, ethyl-4-iodobenzoate, benzoicacid,4-iodo-,ethylester, 4-iodo-benzoic acid ethyl ester, benzoic acid, 4-iodo-, ethyl ester, ethyl4-iodobenzoate, ethyl-4 iodobenzoate, ethyl,4-iodobenzoate
CCOC(=O)C1=CC=C(I)C=C1
276.07
276.08

Chemical Identifiers

51934-41-9
276.07
YCBJOQUNPLTBGG-UHFFFAOYSA-N
142891
CCOC(=O)C1=CC=C(I)C=C1
C9H9IO2
MFCD00017344
p-iodobenzoic acid ethyl ester, 4-iodobenzoic acid ethyl ester, ethyl p-iodobenzoate, ethyl-4-iodobenzoate, benzoicacid,4-iodo-,ethylester, 4-iodo-benzoic acid ethyl ester, benzoic acid, 4-iodo-, ethyl ester, ethyl4-iodobenzoate, ethyl-4 iodobenzoate, ethyl,4-iodobenzoate
ethyl 4-iodobenzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 429-550-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only