missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-iodobenzoate, 98%

Catalog No. p-7021998
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1457009 10 g
AAA1457018 50 g
AAA1457030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1457009 Supplier Thermo Scientific Chemicals Supplier No. A1457009
Only null left

CAS: 51934-41-9 | C9H9IO2 | 276.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51934-41-9
Molecular Formula C9H9IO2
Molecular Weight (g/mol) 276.07
MDL Number MFCD00017344
InChI Key YCBJOQUNPLTBGG-UHFFFAOYSA-N
Synonym p-iodobenzoic acid ethyl ester, 4-iodobenzoic acid ethyl ester, ethyl p-iodobenzoate, ethyl-4-iodobenzoate, benzoicacid,4-iodo-,ethylester, 4-iodo-benzoic acid ethyl ester, benzoic acid, 4-iodo-, ethyl ester, ethyl4-iodobenzoate, ethyl-4 iodobenzoate, ethyl,4-iodobenzoate
PubChem CID 142891
IUPAC Name ethyl 4-iodobenzoate
SMILES CCOC(=O)C1=CC=C(I)C=C1

Specifications

Density 1.66
Boiling Point 283°C
Flash Point >110°C (230°F)
Refractive Index 1.588
Quantity 10 g
Beilstein 2208568
Sensitivity Light sensitive
Formula Weight 276.08
Percent Purity 98%
Chemical Name or Material Ethyl 4-iodobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 429-550-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only