missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-(Chloromethyl)benzoate 98.0+%, TCI America™

Catalog No. E06625G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06625G 5 g
E066225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E06625G Supplier TCI America Supplier No. E06625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1201-90-7
Molecular Formula C10H11ClO2
Molecular Weight (g/mol) 198.646
InChI Key JTTXRFNOFFGPFI-UHFFFAOYSA-N
Synonym 4-(Chloromethyl)benzoic Acid Ethyl Ester
PubChem CID 276519
IUPAC Name ethyl 4-(chloromethyl)benzoate
SMILES CCOC(=O)C1=CC=C(C=C1)CCl

Specifications

Color Colorless
Boiling Point 280°C
Quantity 5 g
UN Number 1760
Formula Weight 198.65
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4-(Chloromethyl)benzoate