accessibility menu, dialog, popup

UserName

CAS RN 1201-90-7

H3COOCl

CAS RN 1201-90-7

IUPAC Name: ethyl 4-(chloromethyl)benzoate
Synonyms: ethyl 4-(chloromethyl)benzoate
Molecular Weight (g/mol): 198.65
Molecular Formula: C10H11ClO2
InChi Key: JTTXRFNOFFGPFI-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(CCl)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results