missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-Bromophenylacetate 98.0+%, TCI America™
$48.13 - $123.21
Chemical Identifiers
| CAS | 14062-25-0 |
|---|---|
| Molecular Formula | C10H11BrO2 |
| Molecular Weight (g/mol) | 243.1 |
| MDL Number | MFCD00016333 |
| InChI Key | ZFDCWHPNBWPPHG-UHFFFAOYSA-N |
| Synonym | ethyl 4-bromophenylacetate, ethyl 2-4-bromophenyl acetate, 4-bromophenylacetic acid ethyl ester, benzeneacetic acid, 4-bromo-, ethyl ester, 4-bromo-phenyl-acetic acid ethyl ester, ethyl 4-bromophenyl acetate, 4-bromo-benzeneacetic acid ethyl ester, ethyl-4-bromophenylacetate, pubchem20039, acmc-209ckw |
| PubChem CID | 7020609 |
| IUPAC Name | ethyl 2-(4-bromophenyl)acetate |
| SMILES | CCOC(=O)CC1=CC=C(C=C1)Br |
Chemical Identifiers
| 14062-25-0 | |
| 243.1 | |
| ZFDCWHPNBWPPHG-UHFFFAOYSA-N | |
| 7020609 | |
| CCOC(=O)CC1=CC=C(C=C1)Br |
| C10H11BrO2 | |
| MFCD00016333 | |
| ethyl 4-bromophenylacetate, ethyl 2-4-bromophenyl acetate, 4-bromophenylacetic acid ethyl ester, benzeneacetic acid, 4-bromo-, ethyl ester, 4-bromo-phenyl-acetic acid ethyl ester, ethyl 4-bromophenyl acetate, 4-bromo-benzeneacetic acid ethyl ester, ethyl-4-bromophenylacetate, pubchem20039, acmc-209ckw | |
| ethyl 2-(4-bromophenyl)acetate |
Specifications
| 14062-25-0 | |
| 144°C | |
| MFCD00016333 | |
| ethyl 4-bromophenylacetate, ethyl 2-4-bromophenyl acetate, 4-bromophenylacetic acid ethyl ester, benzeneacetic acid, 4-bromo-, ethyl ester, 4-bromo-phenyl-acetic acid ethyl ester, ethyl 4-bromophenyl acetate, 4-bromo-benzeneacetic acid ethyl ester, ethyl-4-bromophenylacetate, pubchem20039, acmc-209ckw | |
| CCOC(=O)CC1=CC=C(C=C1)Br | |
| 243.1 | |
| 243.10 | |
| Crystalline Lumps |
| White-Yellow | |
| C10H11BrO2 | |
| 5 g | |
| ZFDCWHPNBWPPHG-UHFFFAOYSA-N | |
| ethyl 2-(4-bromophenyl)acetate | |
| 7020609 | |
| ≥98.0% (GC) | |
| Ethyl 4-Bromophenylacetate |
Safety and Handling
TSCA : No