missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-Bromophenylacetate 98.0+%, TCI America™
Supplier: TCI America E090325G
Specifications
| Ethyl 4-Bromophenylacetate | |
| White-Yellow | |
| C10H11BrO2 | |
| 25 g | |
| ZFDCWHPNBWPPHG-UHFFFAOYSA-N | |
| ethyl 2-(4-bromophenyl)acetate | |
| 7020609 | |
| ≥98.0% (GC) |
| 14062-25-0 | |
| 144°C | |
| MFCD00016333 | |
| ethyl 4-bromophenylacetate, ethyl 2-4-bromophenyl acetate, 4-bromophenylacetic acid ethyl ester, benzeneacetic acid, 4-bromo-, ethyl ester, 4-bromo-phenyl-acetic acid ethyl ester, ethyl 4-bromophenyl acetate, 4-bromo-benzeneacetic acid ethyl ester, ethyl-4-bromophenylacetate, pubchem20039, acmc-209ckw | |
| CCOC(=O)CC1=CC=C(C=C1)Br | |
| 243.1 | |
| 243.10 | |
| Crystalline Lumps |
Chemical Identifiers
| 14062-25-0 | |
| 243.1 | |
| ZFDCWHPNBWPPHG-UHFFFAOYSA-N | |
| 7020609 | |
| CCOC(=O)CC1=CC=C(C=C1)Br |
| C10H11BrO2 | |
| MFCD00016333 | |
| ethyl 4-bromophenylacetate, ethyl 2-4-bromophenyl acetate, 4-bromophenylacetic acid ethyl ester, benzeneacetic acid, 4-bromo-, ethyl ester, 4-bromo-phenyl-acetic acid ethyl ester, ethyl 4-bromophenyl acetate, 4-bromo-benzeneacetic acid ethyl ester, ethyl-4-bromophenylacetate, pubchem20039, acmc-209ckw | |
| ethyl 2-(4-bromophenyl)acetate |
Safety and Handling
TSCA : No