missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-(bromomethyl)benzoate, 96%

Catalog No. AAL1921906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1921906 5 g
AAL1921914 25 g
AAL1921922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1921906 Supplier Thermo Scientific Chemicals Supplier No. L1921906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 26496-94-6 | C10H11BrO2 | 243.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26496-94-6
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.10
MDL Number MFCD03791307
InChI Key TWQLMAJROCNXEA-UHFFFAOYSA-N
Synonym ethyl 4-bromomethyl benzoate, 4-bromomethyl benzoic acid ethyl ester, 4-bromomethyl-benzoic acid ethyl ester, 4-ethoxycarbonylbenzyl bromide, benzoic acid, 4-bromomethyl-, ethyl ester, zlchem 925, acmc-1cknv, 4-carboethoxy-benzylbromide, 4-carboethoxybenzyl bromide, ethyl 4-bromomethyl-benzoate
PubChem CID 954261
IUPAC Name ethyl 4-(bromomethyl)benzoate
SMILES CCOC(=O)C1=CC=C(CBr)C=C1

Specifications

Melting Point 28°C to 33°C
Boiling Point 145°C (5 mmHg)
Refractive Index 1.5494
Quantity 5 g
UN Number UN1759
Beilstein 1869569
Formula Weight 243.1
Percent Purity 96%
Chemical Name or Material Ethyl 4-(bromomethyl)benzoate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only