CAS RN 26496-94-6
IUPAC Name:
ethyl 4-(bromomethyl)benzoate
Synonyms:
ethyl 4-(bromomethyl)benzoate
Molecular Weight (g/mol):
243.1
Molecular Formula:
C10H11BrO2
InChi Key:
TWQLMAJROCNXEA-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC=C(CBr)C=C1
1
–
1
of
1
results
1
Promotions Available
Ethyl 4-(bromomethyl)benzoate, 96%
Ethyl 4-(bromomethyl)benzoate, 96%
CAS: 26496-94-6 Molecular Formula: C10H11BrO2 Molecular Weight (g/mol): 243.10 MDL Number: MFCD03791307 InChI Key: TWQLMAJROCNXEA-UHFFFAOYSA-N Synonym: ethyl 4-bromomethyl benzoate,4-bromomethyl benzoic acid ethyl ester,4-bromomethyl-benzoic acid ethyl ester,4-ethoxycarbonylbenzyl bromide,benzoic acid, 4-bromomethyl-, ethyl ester,zlchem 925,acmc-1cknv,4-carboethoxy-benzylbromide,4-carboethoxybenzyl bromide,ethyl 4-bromomethyl-benzoate PubChem CID: 954261 IUPAC Name: ethyl 4-(bromomethyl)benzoate SMILES: CCOC(=O)C1=CC=C(CBr)C=C1