missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4,4,4-Trifluorocrotonate 98.0+%, TCI America™
Supplier: TCI America E077225G
Specifications
| Ethyl 4,4,4-Trifluorocrotonate | |
| Colorless | |
| C6H7F3O2 | |
| 25 g | |
| ethyl 4,4,4-trifluorocrotonate, ethyl 2e-4,4,4-trifluorobut-2-enoate, ethyl4,4,4-trifluorocrotonate, e-ethyl 4,4,4-trifluorobut-2-enoate, 4,4,4-trifluorocrotonic acid ethyl ester, ethyl e-4,4,4-trifluorobut-2-enoate, ethyl-4,4,4-trifluorocrotonate, ethyl 4,4,4-trifluorobut-2-enoate, 4,4,4-trifluoro-but-2-enoic acid ethyl ester, 2-butenoic acid, 4,4,4-trifluoro-, ethyl ester, 2e | |
| CCOC(=O)\C=C\C(F)(F)F | |
| 168.12 | |
| 168.12 | |
| Liquid |
| 25597-16-4 | |
| 115°C | |
| MFCD00009903 | |
| 3272 | |
| ZKRJCMKLCDWROR-ONEGZZNKSA-N | |
| ethyl (2E)-4,4,4-trifluorobut-2-enoate | |
| 5371261 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 25597-16-4 | |
| 168.12 | |
| ZKRJCMKLCDWROR-ONEGZZNKSA-N | |
| 5371261 | |
| CCOC(=O)\C=C\C(F)(F)F |
| C6H7F3O2 | |
| MFCD00009903 | |
| ethyl 4,4,4-trifluorocrotonate, ethyl 2e-4,4,4-trifluorobut-2-enoate, ethyl4,4,4-trifluorocrotonate, e-ethyl 4,4,4-trifluorobut-2-enoate, 4,4,4-trifluorocrotonic acid ethyl ester, ethyl e-4,4,4-trifluorobut-2-enoate, ethyl-4,4,4-trifluorocrotonate, ethyl 4,4,4-trifluorobut-2-enoate, 4,4,4-trifluoro-but-2-enoic acid ethyl ester, 2-butenoic acid, 4,4,4-trifluoro-, ethyl ester, 2e | |
| ethyl (2E)-4,4,4-trifluorobut-2-enoate |
Safety and Handling
TSCA : No