accessibility menu, dialog, popup

UserName

CH3OOFFF(E)

CAS RN 25597-16-4

IUPAC Name: ethyl (2E)-4,4,4-trifluorobut-2-enoate
Synonyms: ethyl (2E)-4,4,4-trifluorobut-2-enoate
Molecular Weight (g/mol): 168.12
Molecular Formula: C6H7F3O2
InChi Key: ZKRJCMKLCDWROR-ONEGZZNKSA-N
SMILES: CCOC(=O)\C=C\C(F)(F)F
Boiling Point 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)