missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4,4-Trifluorocrotonate 98.0+%, TCI America™

Catalog No. p-7129141
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07725G 5 g
E077225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07725G Supplier TCI America Supplier No. E07725G
Only null left

Chemical Identifiers

CAS 25597-16-4
Molecular Formula C6H7F3O2
Molecular Weight (g/mol) 168.12
MDL Number MFCD00009903
InChI Key ZKRJCMKLCDWROR-ONEGZZNKSA-N
Synonym ethyl 4,4,4-trifluorocrotonate, ethyl 2e-4,4,4-trifluorobut-2-enoate, ethyl4,4,4-trifluorocrotonate, e-ethyl 4,4,4-trifluorobut-2-enoate, 4,4,4-trifluorocrotonic acid ethyl ester, ethyl e-4,4,4-trifluorobut-2-enoate, ethyl-4,4,4-trifluorocrotonate, ethyl 4,4,4-trifluorobut-2-enoate, 4,4,4-trifluoro-but-2-enoic acid ethyl ester, 2-butenoic acid, 4,4,4-trifluoro-, ethyl ester, 2e
PubChem CID 5371261
IUPAC Name ethyl (2E)-4,4,4-trifluorobut-2-enoate
SMILES CCOC(=O)\C=C\C(F)(F)F

Specifications

Color Colorless
Boiling Point 115°C
Quantity 5 g
UN Number 3272
Formula Weight 168.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4,4,4-Trifluorocrotonate