missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(Trifluoromethyl)phenoxyacetate 95.0+%, TCI America™

Catalog No. E10041G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E10041G Supplier TCI America Supplier No. E10041G

Chemical Identifiers

CAS 22897-99-0
Molecular Formula C11H11F3O3
Molecular Weight (g/mol) 248.20
MDL Number MFCD03943910
InChI Key LEKNNERDFCDITP-UHFFFAOYSA-N
Synonym 3-(Trifluoromethyl)phenoxyacetic Acid Ethyl Ester
PubChem CID 4507464
IUPAC Name ethyl 2-[3-(trifluoromethyl)phenoxy]acetate
SMILES CCOC(=O)COC1=CC=CC(=C1)C(F)(F)F

Specifications

Color Yellow
Boiling Point 123°C
Quantity 1 g
Formula Weight 248.20
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-(Trifluoromethyl)phenoxyacetate