CAS RN 22897-99-0
CAS RN 22897-99-0
IUPAC Name:
ethyl 2-[3-(trifluoromethyl)phenoxy]acetate
Synonyms:
ethyl 2-[3-(trifluoromethyl)phenoxy]acetate
Molecular Weight (g/mol):
248.2
Molecular Formula:
C11H11F3O3
InChi Key:
LEKNNERDFCDITP-UHFFFAOYSA-N
SMILES:
CCOC(=O)COC1=CC=CC(=C1)C(F)(F)F
1
–
1
of
1
results
1
Ethyl 3-(Trifluoromethyl)phenoxyacetate 95.0+%, TCI America™
CAS: 22897-99-0 Molecular Formula: C11H11F3O3 Molecular Weight (g/mol): 248.20 MDL Number: MFCD03943910 InChI Key: LEKNNERDFCDITP-UHFFFAOYSA-N Synonym: 3-(Trifluoromethyl)phenoxyacetic Acid Ethyl Ester PubChem CID: 4507464 IUPAC Name: ethyl 2-[3-(trifluoromethyl)phenoxy]acetate SMILES: CCOC(=O)COC1=CC=CC(=C1)C(F)(F)F