missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (3-Hydroxyphenyl)acetate 98.0+%, TCI America™

Catalog No. p-7129410
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12421G 1 g
E12425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12421G Supplier TCI America Supplier No. E12421G
Only null left

Chemical Identifiers

CAS 22446-38-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD07369387
InChI Key NSQBADKMIYCCSC-UHFFFAOYSA-N
Synonym (3-Hydroxyphenyl)acetic Acid Ethyl Ester
PubChem CID 572437
IUPAC Name ethyl 2-(3-hydroxyphenyl)acetate
SMILES CCOC(=O)CC1=CC(O)=CC=C1

Specifications

Boiling Point 179°C
Quantity 1 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl (3-Hydroxyphenyl)acetate