accessibility menu, dialog, popup

UserName

CAS RN 22446-38-4

H3COOOH

CAS RN 22446-38-4

IUPAC Name: ethyl 2-(3-hydroxyphenyl)acetate
Synonyms: ethyl 2-(3-hydroxyphenyl)acetate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: NSQBADKMIYCCSC-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC(O)=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results