missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (3-Hydroxyphenyl)acetate 98.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 179°C |
| Quantity | 5 g |
| Formula Weight | 180.20 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Ethyl (3-Hydroxyphenyl)acetate |
Safety and Handling
| TSCA | No |