Learn More
Ethyl 3-hydroxybenzoate, 99%
CAS: 7781-98-8 | C9H10O3 | 166.176 g/mol
Supplier: Thermo Scientific Chemicals A1534322
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
ApplicationsEthyl 3-hydroxybenzoate has been used in the synthesis of chiral derivatives of ethyl 3-(2-methylbutyloxy)benzoate and 4-(2-methylbutyloxy)-4′-acetylbiphenyl, via Mitsunobu reaction. It is a food-grade antimicrobial agent.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| Ethyl 3-hydroxybenzoate | |
| 72°C to 76°C | |
| C9H10O3 | |
| 100 g | |
| ethyl-3-hydroxybenzoate, benzoic acid, 3-hydroxy-, ethyl ester, 3-hydroxybenzoic acid ethyl ester, m-ethoxycarbonylphenol, ethyl m-hydroxybenzoate, 3-hydroxy-benzoic acid ethyl ester, benzoic acid, m-hydroxy-, ethyl ester, pubchem19923, acmc-209pbw, ethyl 3-oxidanylbenzoate | |
| MWSMNBYIEBRXAL-UHFFFAOYSA-N | |
| ethyl 3-hydroxybenzoate | |
| 24522 | |
| 99% |
| 7781-98-8 | |
| 187°C to 188°C (31 mmHg) | |
| MFCD00002296 | |
| 2208566 | |
| Slightly soluble in water. | |
| CCOC(=O)C1=CC(=CC=C1)O | |
| 166.176 | |
| 166.18 |
Chemical Identifiers
| 7781-98-8 | |
| 166.176 | |
| MWSMNBYIEBRXAL-UHFFFAOYSA-N | |
| 24522 | |
| CCOC(=O)C1=CC(=CC=C1)O |
| C9H10O3 | |
| MFCD00002296 | |
| ethyl-3-hydroxybenzoate, benzoic acid, 3-hydroxy-, ethyl ester, 3-hydroxybenzoic acid ethyl ester, m-ethoxycarbonylphenol, ethyl m-hydroxybenzoate, 3-hydroxy-benzoic acid ethyl ester, benzoic acid, m-hydroxy-, ethyl ester, pubchem19923, acmc-209pbw, ethyl 3-oxidanylbenzoate | |
| ethyl 3-hydroxybenzoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 231-951-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only