missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Cyclopentene-1-carboxylate 97.0+%, TCI America™

Supplier:  TCI America E083125G

 View more versions of this product

Catalog No. E083125G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl 3-Cyclopentene-1-carboxylate
Yellow
C8H12O2
25 g
3-Cyclopentene-1-carboxylic Acid Ethyl Ester
CCOC(=O)C1CC=CC1
140.182
140.18
Liquid
21622-01-5
64°C
MFCD06410819
3272
CTLAIKSGNQPPLO-UHFFFAOYSA-N
ethyl cyclopent-3-ene-1-carboxylate
5314991
≥97.0% (GC)

Chemical Identifiers

21622-01-5
140.182
CTLAIKSGNQPPLO-UHFFFAOYSA-N
5314991
CCOC(=O)C1CC=CC1
C8H12O2
MFCD06410819
3-Cyclopentene-1-carboxylic Acid Ethyl Ester
ethyl cyclopent-3-ene-1-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions