missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Cyclopentene-1-carboxylate 97.0+%, TCI America™

Catalog No. p-7129174
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08315G 5 g
E083125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08315G Supplier TCI America Supplier No. E08315G

Chemical Identifiers

CAS 21622-01-5
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.182
MDL Number MFCD06410819
InChI Key CTLAIKSGNQPPLO-UHFFFAOYSA-N
Synonym 3-Cyclopentene-1-carboxylic Acid Ethyl Ester
PubChem CID 5314991
IUPAC Name ethyl cyclopent-3-ene-1-carboxylate
SMILES CCOC(=O)C1CC=CC1

Specifications

Color Yellow
Boiling Point 64°C
Quantity 5 g
UN Number 3272
Formula Weight 140.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Cyclopentene-1-carboxylate