accessibility menu, dialog, popup

UserName

CAS RN 21622-01-5

H3COO

CAS RN 21622-01-5

IUPAC Name: ethyl cyclopent-3-ene-1-carboxylate
Synonyms: ethyl cyclopent-3-ene-1-carboxylate
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H12O2
InChi Key: CTLAIKSGNQPPLO-UHFFFAOYSA-N
SMILES: CCOC(=O)C1CC=CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results