missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-chlorobenzoate, 98+%

Catalog No. AAL0106514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0106514 25 g
AAL0106506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0106514 Supplier Thermo Scientific Chemicals Supplier No. L0106514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1128-76-3 | C9H9ClO2 | 184.619 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1128-76-3
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00013634
InChI Key LVFRSNCBCHABAM-UHFFFAOYSA-N
Synonym benzoic acid, 3-chloro-, ethyl ester, 3-chlorobenzoic acid ethyl ester, ethyl m-chlorobenzoate, benzoic acid, m-chloro-, ethyl ester, 3-chloro-benzoic acid ethyl ester, pubchem3704, acmc-1bt4u, ethyl3-chlorobenzoate, ksc495m6f, rarechem al bi 0056
PubChem CID 70785
IUPAC Name ethyl 3-chlorobenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)Cl

Specifications

Density 1.186
Boiling Point 118°C to 120°C (13 mmHg)
Refractive Index 1.521
Quantity 25 g
Beilstein 2045877
Formula Weight 184.62
Percent Purity ≥98%
Chemical Name or Material Ethyl 3-chlorobenzoate
EINECSNumber 214-441-3
TSCA Yes

RUO – Research Use Only