missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-Bromopropionate 98.0+%, TCI America™
Supplier: TCI America B0647500G
Specifications
| Ethyl 3-Bromopropionate | |
| Yellow | |
| C5H9BrO2 | |
| 500 g | |
| FQTIYMRSUOADDK-UHFFFAOYSA-N | |
| ethyl 3-bromopropanoate | |
| 68320 | |
| ≥98.0% (GC) |
| 539-74-2 | |
| 136°C | |
| MFCD00000251 | |
| ethyl 3-bromopropionate, propanoic acid, 3-bromo-, ethyl ester, 3-bromopropionic acid ethyl ester, ethyl beta-bromopropionate, unii-9b28g9s1jv, ccris 9051, propionic acid, 3-bromo-, ethyl ester, ethyl .beta.-bromopropionate, 3-bromo-propionic acid ethyl ester, ethyl-3-bromopropionate | |
| CCOC(=O)CCBr | |
| 181.03 | |
| 181.03 | |
| Liquid |
Chemical Identifiers
| 539-74-2 | |
| 181.03 | |
| FQTIYMRSUOADDK-UHFFFAOYSA-N | |
| 68320 | |
| CCOC(=O)CCBr |
| C5H9BrO2 | |
| MFCD00000251 | |
| ethyl 3-bromopropionate, propanoic acid, 3-bromo-, ethyl ester, 3-bromopropionic acid ethyl ester, ethyl beta-bromopropionate, unii-9b28g9s1jv, ccris 9051, propionic acid, 3-bromo-, ethyl ester, ethyl .beta.-bromopropionate, 3-bromo-propionic acid ethyl ester, ethyl-3-bromopropionate | |
| ethyl 3-bromopropanoate |
Safety and Handling
TSCA : Yes