accessibility menu, dialog, popup

UserName

H3COOBr

CAS RN 539-74-2

IUPAC Name: ethyl 3-bromopropanoate
Synonyms: ethyl 3-bromopropanoate
Molecular Weight (g/mol): 181.03
Molecular Formula: C5H9BrO2
InChi Key: FQTIYMRSUOADDK-UHFFFAOYSA-N
SMILES: CCOC(=O)CCBr
Boiling Point 
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)