missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Bromopropionate 98.0+%, TCI America™

Catalog No. B064725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B064725G 25 g
B0647100G 100 g
B0647500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B064725G Supplier TCI America Supplier No. B064725G

Chemical Identifiers

CAS 539-74-2
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00000251
InChI Key FQTIYMRSUOADDK-UHFFFAOYSA-N
Synonym ethyl 3-bromopropionate, propanoic acid, 3-bromo-, ethyl ester, 3-bromopropionic acid ethyl ester, ethyl beta-bromopropionate, unii-9b28g9s1jv, ccris 9051, propionic acid, 3-bromo-, ethyl ester, ethyl .beta.-bromopropionate, 3-bromo-propionic acid ethyl ester, ethyl-3-bromopropionate
PubChem CID 68320
IUPAC Name ethyl 3-bromopropanoate
SMILES CCOC(=O)CCBr

Specifications

Color Yellow
Boiling Point 136°C
Quantity 25 g
Formula Weight 181.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Bromopropionate