missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-aminobenzoate, 98%

Catalog No. AAA1444809
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1444809 10 g
AAA1444818 50 g
AAA1444822 100 g
AAA1444830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1444809 Supplier Thermo Scientific Chemicals Supplier No. A1444809

CAS: 582-33-2 | C9H11NO2 | 165.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 582-33-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00007794
InChI Key ZMCBYSBVJIMENC-UHFFFAOYSA-N
Synonym 3-aminobenzoic acid ethyl ester, m-ethoxycarbonylaniline, 3-ethoxycarbonyl aniline, ethyl m-aminobenzoate, benzoic acid, 3-amino-, ethyl ester, m-aminobenzoic acid, ethyl ester, m-aminobenzoate, metacaine, 3-amino-benzoic acid ethyl ester, benzoic acid, m-amino-, ethyl ester
PubChem CID 11400
ChEBI CHEBI:81494
IUPAC Name ethyl 3-aminobenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)N

Specifications

Melting Point 27°C to 30°C
Density 1.106
Boiling Point 294°C
Flash Point >110°C (230°F)
Refractive Index 1.561
Quantity 10 g
Beilstein 1101967
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material Ethyl 3-aminobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-482-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only