missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,4-Dihydroxybenzoate 98.0+%, TCI America™

Catalog No. D05715G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D05715G 5 g
D057125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D05715G Supplier TCI America Supplier No. D05715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3943-89-3
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00002199
InChI Key KBPUBCVJHFXPOC-UHFFFAOYSA-N
Synonym ethyl protocatechuate, 3,4-dihydroxybenzoic acid ethyl ester, ethyl-3,4-dihydroxybenzoate, unii-4ygj96wtbg, benzoic acid, 3,4-dihydroxy-, ethyl ester, edhb, protocatechuic acid ethyl ester, 4ygj96wtbg, 3,4-dihydroxy-benzoic acid ethyl ester
PubChem CID 77547
IUPAC Name ethyl 3,4-dihydroxybenzoate
SMILES CCOC(=O)C1=CC=C(O)C(O)=C1

Specifications

Melting Point 133°C
Quantity 5 g
Formula Weight 182.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3,4-Dihydroxybenzoate