accessibility menu, dialog, popup

UserName

CAS RN 3943-89-3

IUPAC Name: ethyl 3,4-dihydroxybenzoate
Synonyms: ethyl protocatechuate
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: KBPUBCVJHFXPOC-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(O)C(O)=C1
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)