missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Iodopropionate 97.0+%, TCI America™
$160.30 - $469.75
Chemical Identifiers
| CAS | 31253-08-4 |
|---|---|
| Molecular Formula | C5H9IO2 |
| Molecular Weight (g/mol) | 228.029 |
| MDL Number | MFCD02258628 |
| InChI Key | AVMMXNKUHBWIMU-UHFFFAOYSA-N |
| Synonym | 2-Iodopropionic Acid Ethyl Ester |
| PubChem CID | 10922254 |
| IUPAC Name | ethyl 2-iodopropanoate |
| SMILES | CCOC(=O)C(C)I |
Chemical Identifiers
| 31253-08-4 | |
| 228.029 | |
| AVMMXNKUHBWIMU-UHFFFAOYSA-N | |
| 10922254 | |
| CCOC(=O)C(C)I |
| C5H9IO2 | |
| MFCD02258628 | |
| 2-Iodopropionic Acid Ethyl Ester | |
| ethyl 2-iodopropanoate |
Specifications
| 31253-08-4 | |
| C5H9IO2 | |
| 5 g | |
| AVMMXNKUHBWIMU-UHFFFAOYSA-N | |
| ethyl 2-iodopropanoate | |
| 10922254 | |
| ≥97.0% (GC) | |
| Ethyl 2-Iodopropionate |
| 79°C | |
| MFCD02258628 | |
| 2-Iodopropionic Acid Ethyl Ester | |
| CCOC(=O)C(C)I | |
| 228.029 | |
| 228.03 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Freezer