missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Iodobenzoate 98.0+%, TCI America™

Supplier:  TCI America E127325G

 View more versions of this product

Catalog No. E127325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl 2-Iodobenzoate
165°C
MFCD00051796
2-iodobenzoic acid ethyl ester, rarechem al bi 0562, ethyl iodobenzoate, ethyl-2-iodobenzoate, ethyl 2-iodanylbenzoate, acmc-1ag98, 2-ethoxycarbonyl iodobenzene, benzoic acid, 2-iodo-,ethyl ester, benzoic acid, 2-iodo-, ethyl ester
CCOC(=O)C1=CC=CC=C1I
276.07
276.07
Liquid
1829-28-3
C9H9IO2
25 g
QOUFDDUDXYJWHV-UHFFFAOYSA-N
ethyl 2-iodobenzoate
2829347
≥98.0% (GC)

Chemical Identifiers

1829-28-3
276.07
QOUFDDUDXYJWHV-UHFFFAOYSA-N
2829347
CCOC(=O)C1=CC=CC=C1I
C9H9IO2
MFCD00051796
2-iodobenzoic acid ethyl ester, rarechem al bi 0562, ethyl iodobenzoate, ethyl-2-iodobenzoate, ethyl 2-iodanylbenzoate, acmc-1ag98, 2-ethoxycarbonyl iodobenzene, benzoic acid, 2-iodo-,ethyl ester, benzoic acid, 2-iodo-, ethyl ester
ethyl 2-iodobenzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions