missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Iodobenzoate 98.0+%, TCI America™

Catalog No. p-7129421
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12735G 5 g
E127325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12735G Supplier TCI America Supplier No. E12735G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1829-28-3
Molecular Formula C9H9IO2
Molecular Weight (g/mol) 276.07
MDL Number MFCD00051796
InChI Key QOUFDDUDXYJWHV-UHFFFAOYSA-N
Synonym 2-iodobenzoic acid ethyl ester, rarechem al bi 0562, ethyl iodobenzoate, ethyl-2-iodobenzoate, ethyl 2-iodanylbenzoate, acmc-1ag98, 2-ethoxycarbonyl iodobenzene, benzoic acid, 2-iodo-,ethyl ester, benzoic acid, 2-iodo-, ethyl ester
PubChem CID 2829347
IUPAC Name ethyl 2-iodobenzoate
SMILES CCOC(=O)C1=CC=CC=C1I

Specifications

Boiling Point 165°C
Quantity 5 g
Formula Weight 276.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Iodobenzoate