missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Hydroxyisobutyrate 98.0+%, TCI America™
Supplier: TCI America H0255250G
Specifications
| Ethyl 2-Hydroxyisobutyrate | |
| Colorless | |
| C6H12O3 | |
| 250 g | |
| ethyl 2-hydroxyisobutyrate, ethyl-2-hydroxyisobutyrate, ethyl 2-hydroxy-2-methylpropionate, propanoic acid, 2-hydroxy-2-methyl-, ethyl ester, propanoic acid, 2-hydroxy-2-methyl-, ethyl ester, 2-methyllactic acid ethyl ester, ethyl, ethyl, a-hydroxyisobutyrate, unii-s1530mcv3x, unii-s1530mcv3x | |
| CCOC(=O)C(C)(C)O | |
| 132.16 | |
| 132.16 | |
| Liquid |
| 80-55-7 | |
| 150°C | |
| MFCD00004458 | |
| 3272 | |
| GFUIDHWFLMPAGY-UHFFFAOYSA-N | |
| ethyl 2-hydroxy-2-methylpropanoate | |
| 6653 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 80-55-7 | |
| 132.16 | |
| GFUIDHWFLMPAGY-UHFFFAOYSA-N | |
| 6653 | |
| CCOC(=O)C(C)(C)O |
| C6H12O3 | |
| MFCD00004458 | |
| ethyl 2-hydroxyisobutyrate, ethyl-2-hydroxyisobutyrate, ethyl 2-hydroxy-2-methylpropionate, propanoic acid, 2-hydroxy-2-methyl-, ethyl ester, propanoic acid, 2-hydroxy-2-methyl-, ethyl ester, 2-methyllactic acid ethyl ester, ethyl, ethyl, a-hydroxyisobutyrate, unii-s1530mcv3x, unii-s1530mcv3x | |
| ethyl 2-hydroxy-2-methylpropanoate |
Safety and Handling
EINECSNumber : (2)-1430
RTECSNumber : OD5672000
TSCA : Yes