missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Hydroxyisobutyrate 98.0+%, TCI America™

Catalog No. p-7130445
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H025525G 25 g
H0255250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H025525G Supplier TCI America Supplier No. H025525G
Only null left

Chemical Identifiers

CAS 80-55-7
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00004458
InChI Key GFUIDHWFLMPAGY-UHFFFAOYSA-N
Synonym ethyl 2-hydroxyisobutyrate, ethyl 2-hydroxy-2-methylpropionate, ethyl 2-methyllactate, propanoic acid, 2-hydroxy-2-methyl-, ethyl ester, ethyl alpha-hydroxyisobutyrate, 2-methyllactic acid ethyl ester, ethyl, a-hydroxyisobutyrate, lactic acid, 2-methyl-, ethyl ester, unii-s1530mcv3x, ethyl-2-hydroxyisobutyrate
PubChem CID 6653
IUPAC Name ethyl 2-hydroxy-2-methylpropanoate
SMILES CCOC(=O)C(C)(C)O

Specifications

Color Colorless
Boiling Point 150°C
Quantity 25 g
UN Number 3272
Formula Weight 132.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Hydroxyisobutyrate
EINECSNumber (2)-1430
RTECSNumber OD5672000
TSCA Yes