missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-fluorophenylacetate, 99%, Thermo Scientific Chemicals

Catalog No. AAL1999906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1999906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1999906 Supplier Thermo Scientific Chemicals Supplier No. L1999906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 584-74-7 | Molecular Formula: C10H11FO2 | Molecular Weight: 182.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 584-74-7
Molecular Formula C10H11FO2
Molecular Weight (g/mol) 182.194
MDL Number MFCD04039343
InChI Key OFWMNCYZASDVSS-UHFFFAOYSA-N
Synonym ethyl 2-2-fluorophenyl acetate, ethyl 2-fluorophenylacetate, ethyl 2-fluorophenyl acetate, 2-fluorophenylacetic acid ethyl ester, benzeneacetic acid, 2-fluoro-, ethyl ester, acmc-209uz7, ethyl 2-fluorobenzeneacetate
PubChem CID 7578179
IUPAC Name ethyl 2-(2-fluorophenyl)acetate
SMILES CCOC(=O)CC1=CC=CC=C1F

Specifications

Density 1.101
Boiling Point 235°C
Flash Point 85°C (185°F)
Refractive Index 1.482
Quantity 5 g
Formula Weight 182.2
Percent Purity 99%
Chemical Name or Material Ethyl 2-fluorophenylacetate
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only