CAS RN 584-74-7
IUPAC Name:
ethyl 2-(2-fluorophenyl)acetate
Synonyms:
ethyl 2-(2-fluorophenyl)acetate
Molecular Weight (g/mol):
182.19
Molecular Formula:
C10H11FO2
InChi Key:
OFWMNCYZASDVSS-UHFFFAOYSA-N
SMILES:
CCOC(=O)CC1=CC=CC=C1F
1
–
1
of
1
results
1
Promotions Available
Ethyl 2-fluorophenylacetate, 99%, Thermo Scientific Chemicals
Ethyl 2-fluorophenylacetate, 99%, Thermo Scientific Chemicals
CAS: 584-74-7 Molecular Formula: C10H11FO2 Molecular Weight (g/mol): 182.194 MDL Number: MFCD04039343 InChI Key: OFWMNCYZASDVSS-UHFFFAOYSA-N Synonym: ethyl 2-2-fluorophenyl acetate,ethyl 2-fluorophenylacetate,ethyl 2-fluorophenyl acetate,2-fluorophenylacetic acid ethyl ester,benzeneacetic acid, 2-fluoro-, ethyl ester,acmc-209uz7,ethyl 2-fluorobenzeneacetate PubChem CID: 7578179 IUPAC Name: ethyl 2-(2-fluorophenyl)acetate SMILES: CCOC(=O)CC1=CC=CC=C1F