missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Fluorobenzoate 98.0+%, TCI America™

Catalog No. p-7129660
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03955G 5 g
F039525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03955G Supplier TCI America Supplier No. F03955G

Chemical Identifiers

CAS 443-26-5
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.167
MDL Number MFCD00039214
InChI Key RUWPGPOBTHOLHF-UHFFFAOYSA-N
Synonym ethyl o-fluorobenzoate, 2-fluorobenzoic acid ethyl ester, benzoic acid, 2-fluoro-, ethyl ester, ethyl2-fluorobenzoate, benzoic acid, o-fluoro-, ethyl ester, 2-fluoro-benzoic acid ethyl ester, pubchem3469, ethyl-2-fluorobenzoate, ethyl 2-fluoro-benzoate, acmc-1an49
PubChem CID 67953
IUPAC Name ethyl 2-fluorobenzoate
SMILES CCOC(=O)C1=CC=CC=C1F

Specifications

Color Yellow
Boiling Point 221°C
Quantity 5 g
Formula Weight 168.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Fluorobenzoate