CAS RN 443-26-5
IUPAC Name:
ethyl 2-fluorobenzoate
Synonyms:
ethyl 2-fluorobenzoate
Molecular Weight (g/mol):
168.17
Molecular Formula:
C9H9FO2
InChi Key:
RUWPGPOBTHOLHF-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC=CC=C1F
1
–
1
of
1
results
1
Ethyl 2-Fluorobenzoate 98.0+%, TCI America™
CAS: 443-26-5 Molecular Formula: C9H9FO2 Molecular Weight (g/mol): 168.167 MDL Number: MFCD00039214 InChI Key: RUWPGPOBTHOLHF-UHFFFAOYSA-N Synonym: ethyl o-fluorobenzoate,2-fluorobenzoic acid ethyl ester,benzoic acid, 2-fluoro-, ethyl ester,ethyl2-fluorobenzoate,benzoic acid, o-fluoro-, ethyl ester,2-fluoro-benzoic acid ethyl ester,pubchem3469,ethyl-2-fluorobenzoate,ethyl 2-fluoro-benzoate,acmc-1an49 PubChem CID: 67953 IUPAC Name: ethyl 2-fluorobenzoate SMILES: CCOC(=O)C1=CC=CC=C1F