missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Ethoxybenzoate 98.0+%, TCI America™

Catalog No. E041325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E041325G 25 g
E0413500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E041325G Supplier TCI America Supplier No. E041325G

Chemical Identifiers

CAS 6290-24-0
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00142907
InChI Key OUZCDRGUTZLAGO-UHFFFAOYSA-N
Synonym 2-Ethoxybenzoic Acid Ethyl Ester
PubChem CID 221644
IUPAC Name ethyl 2-ethoxybenzoate
SMILES CCOC1=CC=CC=C1C(=O)OCC

Specifications

Color Yellow
Boiling Point 251°C
Quantity 25 g
Formula Weight 194.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Ethoxybenzoate