accessibility menu, dialog, popup

UserName

CAS RN 6290-24-0

CH3OOOCH3

CAS RN 6290-24-0

IUPAC Name: ethyl 2-ethoxybenzoate
Synonyms: ethyl 2-ethoxybenzoate
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: OUZCDRGUTZLAGO-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC=C1OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results