missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Cyano-3,3-diphenylacrylate 98.0+%, TCI America™
$52.19 - $369.08
Chemical Identifiers
| CAS | 5232-99-5 |
|---|---|
| Molecular Formula | C18H15NO2 |
| Molecular Weight (g/mol) | 277.323 |
| MDL Number | MFCD00027364 |
| InChI Key | IAJNXBNRYMEYAZ-UHFFFAOYSA-N |
| Synonym | 2-Cyano-3,3-diphenylacrylic Acid Ethyl Ester |
| PubChem CID | 243274 |
| IUPAC Name | ethyl 2-cyano-3,3-diphenylprop-2-enoate |
| SMILES | CCOC(=O)C(=C(C1=CC=CC=C1)C2=CC=CC=C2)C#N |
Chemical Identifiers
| 5232-99-5 | |
| 277.323 | |
| IAJNXBNRYMEYAZ-UHFFFAOYSA-N | |
| 243274 | |
| CCOC(=O)C(=C(C1=CC=CC=C1)C2=CC=CC=C2)C#N |
| C18H15NO2 | |
| MFCD00027364 | |
| 2-Cyano-3,3-diphenylacrylic Acid Ethyl Ester | |
| ethyl 2-cyano-3,3-diphenylprop-2-enoate |
Specifications
| 5232-99-5 | |
| White-Yellow | |
| C18H15NO2 | |
| 25 g | |
| IAJNXBNRYMEYAZ-UHFFFAOYSA-N | |
| ethyl 2-cyano-3,3-diphenylprop-2-enoate | |
| 243274 | |
| ≥98.0% (GC) | |
| Ethyl 2-Cyano-3,3-diphenylacrylate |
| 98°C | |
| 200°C | |
| MFCD00027364 | |
| 2-Cyano-3,3-diphenylacrylic Acid Ethyl Ester | |
| CCOC(=O)C(=C(C1=CC=CC=C1)C2=CC=CC=C2)C#N | |
| 277.323 | |
| 277.32 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-2792&(4)-0262
RTECSNumber : AT6200000
TSCA : Yes