accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 5232-99-5

H3COON

CAS RN 5232-99-5

IUPAC Name: ethyl 2-cyano-3,3-diphenylprop-2-enoate
Synonyms: etocrylene uvinul N 35 etocrilene etocrileno etocrilenum UV absorber-2
Molecular Weight (g/mol): 277.32
Molecular Formula: C18H15NO2
InChi Key: IAJNXBNRYMEYAZ-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C#N)=C(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.