missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromophenylacetate, 98%

Catalog No. p-7033507
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50630MD 250 mg
AAH5063003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50630MD Supplier Thermo Scientific Chemicals Supplier No. H50630MD

CAS: 2178-24-7 | C10H11BrO2 | 243.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2178-24-7
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.1
MDL Number MFCD00051921
InChI Key MZQXAVZPEZUJIJ-UHFFFAOYSA-N
Synonym ethyl 2-2-bromophenyl acetate, ethyl 2-bromophenylacetate, ethyl 2-bromophenyl acetate, benzeneacetic acid, 2-bromo-, ethyl ester, acmc-20al1y, 2-bromo-phenyl-acetic acid ethyl ester, ethyl o-bromophenylacetate, ethyl-2-bromophenylacetate, ethyl 2-bromo-phenylacetate, ethyl 2-2-bromophenyl-acetate
PubChem CID 2780092
IUPAC Name ethyl 2-(2-bromophenyl)acetate
SMILES CCOC(=O)CC1=CC=CC=C1Br

Specifications

Melting Point 35°C to 38°C
Quantity 250 mg
Formula Weight 243.1
Percent Purity 98%
Chemical Name or Material Ethyl 2-bromophenylacetate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only